Learn More
Strem, An Ascensus Company CAS# [352535-01-4]. 2g. Tetrakis(ethylmethylamino)hafnium, 99% (99.99+%-Hf, <0.15% Zr) TEMAH PURATREM. MFCD03427130
SDP
Supplier:  Strem, An Ascensus Company 7277202G
Encompass
CAS# 352535-01-4. 2g. Tetrakis(ethylmethylamino)hafnium, 99% (99.99+%-Hf, <0.15% Zr) TEMAH PURATREM. MFCD03427130. Molecular Weight: 410.90. Molecular Formula: Hf[N(CH3)(CH2CH3)]4. Color/form: colorless to yellow liq. Strem# 72-7720. www.strem.com/catalog/v3/72-7720/
Catalog No. 50-197-9352