Learn More
Medchemexpress LLC HY-10560 1mg Medchemexpress, Temanogrel CAS:887936-68-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105601MG
Encompass_Preferred
Medchemexpress, HY-10560 1mg Temanogrel CAS:887936-68-7 Temanogrel is a highly selective 5-HT2A receptor antagonist with a Ki of 4.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8279
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.