Learn More
Medchemexpress LLC Temanogrel | 887936-68-7 | 98.9% | 436.50 g·mol⁻¹ | C24H28N4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1056025MG
Encompass_Preferred
Temanogrel is a research-grade selective 5-HT2A receptor antagonist (reported Ki 4.9 nM) used to study serotonin receptor pharmacology and platelet function. Supplied as a solid for laboratory research use only, it is provided with stability information for short- and long-term storage.
  • Selective 5-HT2A receptor antagonist (Ki 4.9 nM).
  • High purity (98.9%).
  • Solid form with white to light yellow appearance.
  • Recommended storage conditions provided for solid and solution stability.
  • Suitable for in vitro receptor binding and platelet function studies.
Catalog No. 50-002-40679