Learn More
Medchemexpress LLC Temanogrel | 887936-68-7 | 98.9% | 436.50 g·mol⁻¹ | C24H28N4O4 | 50 MG
SDP
Catalog No. 5000241142
Encompass_Preferred
Temanogrel is a selective 5-HT2A receptor antagonist used as a research compound to study serotonin-mediated platelet aggregation and cardiovascular pharmacology. It is supplied as a high-purity solid intended for laboratory research use only and requires low-temperature storage for long-term stability.
  • Selective 5-HT2A receptor antagonist with Ki 4.9 nM.
  • High purity suitable for analytical and biological assays.
  • Supplied as a solid with recommended low-temperature storage.
  • Stable in powder form for extended storage at -20°C.
Catalog No. 50-002-41142 Supplier Medchemexpress LLC Supplier No. HY1056050MG
Temanogrel is a selective 5-HT2A receptor antagonist used as a research compound to study serotonin-mediated platelet aggregation and cardiovascular pharmacology. It is supplied as a high-purity solid intended for laboratory research use only and requires low-temperature storage for long-term stability.
  • Selective 5-HT2A receptor antagonist with Ki 4.9 nM.
  • High purity suitable for analytical and biological assays.
  • Supplied as a solid with recommended low-temperature storage.
  • Stable in powder form for extended storage at -20°C.