Learn More
Strem, An Ascensus Company CAS# 175923-04-3. 1g. Tetrakis(ethylmethylamino)zirconium(IV), 99% TEMAZ. MFCD03427131
SDP
Supplier:  Strem, An Ascensus Company 4017101G
Encompass
CAS# 175923-04-3. 1g. Tetrakis(ethylmethylamino)zirconium(IV), 99% TEMAZ. MFCD03427131. Molecular Weight: 323.63. Molecular Formula: Zr[N(CH3)(CH2CH3)]4. Color/form: light yellow liq. Strem# 40-1710. http://www.strem.com/catalog/v/40-1710/
Catalog No. 50-204-5305