Learn More
Biotang Inc TEMED, 10 ML, P-1520, CAS # 110-18-9. (CH3)2NCH2CH2N(CH3)2 with mol. wt. of 116.20. Liquid with purity of > 99%, and density of 0.775 g/ml (at 20 degrees Celsius).
Shop All BIOTANG Inc Products