Learn More
AG Scientific Inc TEMED 100 ML
Catalog No. 502268400
Encompass_Preferred
TEMED (N,N,N',N'-Tetramethylethylenediamine) is a tertiary amine used to catalyze formation of free radicals from ammonium persulfate (APS) and riboflavin. Because of its properties, TEMED promotes the acrylamide polymerization process.TEMED is suitable for electrophoresis and has been tested as a catalyst for polyacrylamide gel electrophoresis. However, it may affect the characteristics of sample proteins and the molecular weight measured, depending on the concentration of TEMED used for the experiment.
Catalog No. 50-226-8400 Supplier AG Scientific Inc Supplier No. T2699100ML
TEMED (N,N,N',N'-Tetramethylethylenediamine) is a tertiary amine used to catalyze formation of free radicals from ammonium persulfate (APS) and riboflavin. Because of its properties, TEMED promotes the acrylamide polymerization process.TEMED is suitable for electrophoresis and has been tested as a catalyst for polyacrylamide gel electrophoresis. However, it may affect the characteristics of sample proteins and the molecular weight measured, depending on the concentration of TEMED used for the experiment.