Learn More
Abcam TEMED, free radical stabilizer, 25ML
Catalog No. 502704814
Encompass_Preferred
MW 116.21 g/mol, Purity >99%. Tertiary amine base used to catalyze the formation of free radicals from ammonium persulfate (APS) or riboflavin. The free radicals will cause acrylamide and bis-acrylamide to polymerize to form a gel matrix which can be used for sieving macromolecules such as nucleic acids and proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4814 Supplier Abcam Supplier No. AB14623525ML
MW 116.21 g/mol, Purity >99%. Tertiary amine base used to catalyze the formation of free radicals from ammonium persulfate (APS) or riboflavin. The free radicals will cause acrylamide and bis-acrylamide to polymerize to form a gel matrix which can be used for sieving macromolecules such as nucleic acids and proteins.
The product is subject to the following: Abcam Restricted Use Statement