Learn More
National Diagnostics TEMED - ULTA PURE 25 ML
SDP
Catalog No. 5089990038
Encompass
TEMED - ULTA PURE, Fractionally distilled to remove all trace metals and amine impurities. Stored under nitrogen, 25 ML
Catalog No. 50-899-90038 Supplier National Diagnostics Supplier No. EC50325ML
TEMED - ULTA PURE, Fractionally distilled to remove all trace metals and amine impurities. Stored under nitrogen, 25 ML