Learn More
Cayman Chemical Temocapril (hydrochloride), Purity: ≥98%, CAS: 110221-44-8, 10 mg
SDP
Catalog No. 502966037
Encompass_Preferred
A prodrug form of temocaprilat; prevents impairment of acetylcholine-induced relaxation in aortic rings isolated from diabetic rats in a model of streptozotocin-induced diabetes; inhibits the angiotensin I-induced pressor response in normotensive rats at 0.1, 0.3, and 1 mg/kg; prevents coronary vascular remodeling induced by L-NAME in rats at 0.1 mg/ml in the drinking water; reduces plasma glucose levels in a KKAy mouse model of type 2 diabetes at 1 mg/kg
Catalog No. 50-296-6037 Supplier Cayman Chemical Supplier No. 1824110 MG
A prodrug form of temocaprilat; prevents impairment of acetylcholine-induced relaxation in aortic rings isolated from diabetic rats in a model of streptozotocin-induced diabetes; inhibits the angiotensin I-induced pressor response in normotensive rats at 0.1, 0.3, and 1 mg/kg; prevents coronary vascular remodeling induced by L-NAME in rats at 0.1 mg/ml in the drinking water; reduces plasma glucose levels in a KKAy mouse model of type 2 diabetes at 1 mg/kg