Learn More
Medchemexpress LLC Temocillin (disodium) | 61545-06-0 | 92.3% | 458.42 | 100 MG
SDP
Catalog No. 5000351538
Encompass_Preferred
Temocillin disodium is a 6-α-methoxy penicillin that exhibits antibacterial activity. It is intended for research use only and not for patient use.
  • 6-α-Methoxy penicillin structure
  • Possesses antibacterial activity
  • Used in research related to antibacterial agents
  • Demonstrated effectiveness in reducing bacterial titres in kidneys against Escherichia coli in a murine model of urinary tract infection
Catalog No. 50-003-51538 Supplier Medchemexpress LLC Supplier No. HY139597100MG
Temocillin disodium is a 6-α-methoxy penicillin that exhibits antibacterial activity. It is intended for research use only and not for patient use.
  • 6-α-Methoxy penicillin structure
  • Possesses antibacterial activity
  • Used in research related to antibacterial agents
  • Demonstrated effectiveness in reducing bacterial titres in kidneys against Escherichia coli in a murine model of urinary tract infection