Learn More
Medchemexpress LLC Temocillin disodium | 61545-06-0 | 92.3% | 458.42 | C16H16N2Na2O7S2 | 5 MG
SDP
Catalog No. 5000221469
Encompass_Preferred
Temocillin disodium is the disodium salt of temocillin, a 6-α-methoxy penicillin antibiotic provided for research use. It is used in microbiology and antibacterial testing against β-lactamase-producing Gram-negative bacteria and is supplied as a characterized, research-grade chemical.
  • Water-soluble disodium salt suitable for aqueous formulations.
  • Active against β-lactamase-producing Gram-negative bacteria.
  • Available in small research-grade quantities for laboratory studies.
  • Characterized by CAS 61545-06-0 and molecular weight 458.42 for traceability.
  • Supplied with product and safety documentation for laboratory handling.
Catalog No. 50-002-21469 Supplier Medchemexpress LLC Supplier No. HY1395975MG
Temocillin disodium is the disodium salt of temocillin, a 6-α-methoxy penicillin antibiotic provided for research use. It is used in microbiology and antibacterial testing against β-lactamase-producing Gram-negative bacteria and is supplied as a characterized, research-grade chemical.
  • Water-soluble disodium salt suitable for aqueous formulations.
  • Active against β-lactamase-producing Gram-negative bacteria.
  • Available in small research-grade quantities for laboratory studies.
  • Characterized by CAS 61545-06-0 and molecular weight 458.42 for traceability.
  • Supplied with product and safety documentation for laboratory handling.