Learn More
Medchemexpress LLC Temocillin disodium | 61545-06-0 | 92.3% | C16H16N2Na2O7S2 | 10MG
SDP
Catalog No. 5000260944
Encompass_Preferred
Temocillin disodium is the disodium salt form of temocillin, a 6-α-methoxy penicillin supplied as a research reagent and analytical standard. It exhibits antibacterial activity and is intended for laboratory research use only, not for human or clinical applications. Typical specifications include molecular formula C16H16N2Na2O7S2, molecular weight 458.42, and reported purity around 92.3%.
  • Contains a 6-α-methoxy penicillin core with antibacterial activity.
  • Provided as the disodium salt to enhance aqueous solubility.
  • Typical purity approximately 92.3% suitable for research purposes.
  • Characterized by molecular formula C16H16N2Na2O7S2 and molecular weight 458.42.
  • Intended for laboratory research use only; not for human use.
Catalog No. 50-002-60944 Supplier Medchemexpress LLC Supplier No. HY13959710MG
Temocillin disodium is the disodium salt form of temocillin, a 6-α-methoxy penicillin supplied as a research reagent and analytical standard. It exhibits antibacterial activity and is intended for laboratory research use only, not for human or clinical applications. Typical specifications include molecular formula C16H16N2Na2O7S2, molecular weight 458.42, and reported purity around 92.3%.
  • Contains a 6-α-methoxy penicillin core with antibacterial activity.
  • Provided as the disodium salt to enhance aqueous solubility.
  • Typical purity approximately 92.3% suitable for research purposes.
  • Characterized by molecular formula C16H16N2Na2O7S2 and molecular weight 458.42.
  • Intended for laboratory research use only; not for human use.