Learn More
Medchemexpress LLC HY-17364 100mg , Temozolomide CAS:85622-93-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17364100MG
Encompass_Preferred
HY-17364 100mg Temozolomide CAS: 85622-93-1 Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. Temozolomide is also a proautophagic and proapoptotic agent. Temozolomide is effective against tumor cells that are characterized by low levels of O6-alkylguanine DNA alkyltransferase (OGAT) and a functional mismatch repair system. Temozolomide has antitumor and antiangiogenic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9152