Learn More
Cayman Chemical Temozolomide, Purity: ≥98%, CAS: 85622-93-1, 100 mg
SDP
Catalog No. 502953723
Encompass_Preferred
A prodrug form of MTIC; selectively cytotoxic to MGMT-deficient U87 and D54 cells over MGMT-expressing T98G and U3054MG cells (IC50s = 51, 12, 660, and 370 µM, respectively); increases survival in a Br23c glioblastoma orthotopic mouse xenograft model at 15 mg/kg; decreases tumor volume and increases the number of tumor-infiltrating CD4+ and CD8+ T cells in a GL261 murine glioma model in combination with anti-CD47
Catalog No. 50-295-3723 Supplier Cayman Chemical Supplier No. 14163100 MG
A prodrug form of MTIC; selectively cytotoxic to MGMT-deficient U87 and D54 cells over MGMT-expressing T98G and U3054MG cells (IC50s = 51, 12, 660, and 370 µM, respectively); increases survival in a Br23c glioblastoma orthotopic mouse xenograft model at 15 mg/kg; decreases tumor volume and increases the number of tumor-infiltrating CD4+ and CD8+ T cells in a GL261 murine glioma model in combination with anti-CD47