Learn More
Apexbio Technology LLC Temozolomide, 85622-93-1, MFCD00866492, 10 mg
SDP
Supplier:  Apexbio Technology LLC B139910
Encompass_Preferred
MF: C6H6N6O2, MW: 194.15, Solubility: 29.61mg/mL in DMSO. Temozolomide, an alkylating agent, is a DNA damage inducer.
Catalog No. 50-101-1494