Learn More
Enzo Life Sciences Temozolomide (500mg). CAS: 85622-93-1
SDP
Supplier:  Enzo Life Sciences ALX420044M500
Encompass_Preferred
Antitumor and antiangiogenic compound. DNA alkylating agent. Induces G2/M arrest and apoptosis through adduction of a methyl group to O6 position of guanine in genomic DNA. Inactivates DNA repair protein O(6)-alkylguanine DNA alkyltransferase (AGT) in base excision repair (BER) pathway. Chemotherapeutic agent. Modifies drug resistance. Alternative name: 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3H)-one. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO; slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9123