Learn More
Medchemexpress LLC Temozolomide acid | 113942-30-6 | MFCD12032160 | 99.7% | 195.14 g·mol⁻¹ | C6H5N5O3 | 1 G
SDP
Catalog No. 5000300874
Encompass_Preferred
Temozolomide acid is the carboxylic acid derivative and active metabolite of temozolomide used in anticancer research. It mediates cytotoxicity through DNA alkylation-related mechanisms and is commonly used in glioblastoma and DNA damage response studies. Available in solid and solution formats for laboratory use.
  • Used in research of glioblastoma and brain cancer.
  • Acts via DNA alkylation, causing DNA strand breaks and cell death.
  • High purity: 99.7%.
  • Molecular formula C6H5N5O3, molecular weight 195.14 g·mol⁻¹.
  • Available as solid and DMSO solution formats and multiple pack sizes.
  • Safety data sheet available for laboratory handling.
Catalog No. 50-003-00874 Supplier Medchemexpress LLC Supplier No. HYW0183261G
Add to Cart
Add to Cart
Temozolomide acid is the carboxylic acid derivative and active metabolite of temozolomide used in anticancer research. It mediates cytotoxicity through DNA alkylation-related mechanisms and is commonly used in glioblastoma and DNA damage response studies. Available in solid and solution formats for laboratory use.
  • Used in research of glioblastoma and brain cancer.
  • Acts via DNA alkylation, causing DNA strand breaks and cell death.
  • High purity: 99.7%.
  • Molecular formula C6H5N5O3, molecular weight 195.14 g·mol⁻¹.
  • Available as solid and DMSO solution formats and multiple pack sizes.
  • Safety data sheet available for laboratory handling.