Learn More
Medchemexpress LLC Temozolomide-d3 | 208107-14-6 | 99.8% | 197.17 | 1 MG
SDP
Catalog No. 5000384688
Encompass_Preferred
Temozolomide-d3 is the deuterium labeled Temozolomide. Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. It is also a proautophagic and proapoptotic agent. Temozolomide is effective against tumor cells characterized by low levels of O6-alkylguanine DNA alkyltransferase (OGAT) and a functional mismatch repair system. It has antitumor and antiangiogenic effects.
  • Deuterium labeled Temozolomide.
  • Oral active DNA alkylating agent.
  • Crosses the blood-brain barrier.
  • Proautophagic and proapoptotic agent.
  • Effective against tumor cells with low levels of OGAT and a functional mismatch repair system.
  • Antitumor and antiangiogenic effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84688 Supplier Medchemexpress LLC Supplier No. HY17364S1MG
Temozolomide-d3 is the deuterium labeled Temozolomide. Temozolomide (NSC 362856) is an oral active DNA alkylating agent that crosses the blood-brain barrier. It is also a proautophagic and proapoptotic agent. Temozolomide is effective against tumor cells characterized by low levels of O6-alkylguanine DNA alkyltransferase (OGAT) and a functional mismatch repair system. It has antitumor and antiangiogenic effects.
  • Deuterium labeled Temozolomide.
  • Oral active DNA alkylating agent.
  • Crosses the blood-brain barrier.
  • Proautophagic and proapoptotic agent.
  • Effective against tumor cells with low levels of OGAT and a functional mismatch repair system.
  • Antitumor and antiangiogenic effects.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.