Learn More
Hampshire Controls TEMP. ALARMS MODEL 140-100HBR
Catalog No. NC0418310
Encompass_Preferred
Programmable, temp. alarm operates from -100 to +100 deg.C. with high/low temp. alarms, audible/visual alarms, alarm/relay delay periods, battery back-up and relay output. Used for monitoring freezers, refrigerators, walk-in coolers and server rooms.
Catalog No. NC0418310 Supplier Hampshire Controls Supplier No. 140100HRB
Programmable, temp. alarm operates from -100 to +100 deg.C. with high/low temp. alarms, audible/visual alarms, alarm/relay delay periods, battery back-up and relay output. Used for monitoring freezers, refrigerators, walk-in coolers and server rooms.