Learn More
World Precision Instrument Temp Control Box 1 w/cable, Premium Stereotaxic
SDP
Catalog No. 502539083
Encompass
Minimize heat loss and improve surgical outcomes with Rodent Warmer.
  • Easy to use dial control 
  • Preprogrammed animal temperatures
  • Use with or without rectal probe (rectal probe sold seperately)
  • Lightweight, small footprint
  • Controller used exclusively with the Premium Stereotaxic frame
Catalog No. 50-253-9083 Supplier World Precision Instrument Supplier No. 505375
Minimize heat loss and improve surgical outcomes with Rodent Warmer.
  • Easy to use dial control 
  • Preprogrammed animal temperatures
  • Use with or without rectal probe (rectal probe sold seperately)
  • Lightweight, small footprint
  • Controller used exclusively with the Premium Stereotaxic frame