Learn More
Memmert Memmert water bath WTB 35, 115volt +5C above ambient to +100C (Dependant on Ambient) Gable Cover/ Reversible Shelf included
SDP
Catalog No. 5000451098
Encompass_Preferred
Memmert water bath WTB 35, 115volt +5C above ambient to +100C (Dependant on Ambient) Gable Cover/ Reversible Shelf included
Catalog No. 50-004-51098 Supplier Memmert Supplier No. WTB 35 115V
Memmert water bath WTB 35, 115volt +5C above ambient to +100C (Dependant on Ambient) Gable Cover/ Reversible Shelf included