Learn More
Memmert Memmert water bath WTB 11, 115volt +5C above ambient to +100C (Dependant on Ambient) Gable Cover/ Reversible Shelf included
SDP
Catalog No. 5000451117
Encompass_Preferred
Memmert water bath WTB 11, 115volt +5C above ambient to +100C (Dependant on Ambient) Gable Cover/ Reversible Shelf included
Catalog No. 50-004-51117 Supplier Memmert Supplier No. WTB 11 115V
Memmert water bath WTB 11, 115volt +5C above ambient to +100C (Dependant on Ambient) Gable Cover/ Reversible Shelf included