Learn More
Cole-Parmer Environmental Express HotBlock 200 SC1831 Programmable Temperature Controller, Single Block; 1/Ea
Supplier:  Cole-Parmer 5320600
Encompass
Perform a wider range of experiments and digestions with the expanded temperature rangeup to 200C* with the new HotBlock 200! Control one or two block heaters. Choose your controller, then block (or blocks) to suit your metals digestion needs. Mount controllers outside the fume hood on metal surfaces, in an available outlet, or place on a benchtop.
Catalog No. NC1508524