Learn More
Chemglass Life Sciences Dual Temperature Controller Only, J-Kem, Apollo, Type "RTD" (-200°C to 400°C)
SDP
Catalog No. 501311221
Encompass_Preferred
Dual Temperature Controller Only, J-Kem, Apollo, Type "RTD" (-200C to 400C). J-Kem Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet. Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. Temperature ramping and serial communications are also provided with this unit.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 4" H x 8" W x 9" DPower: 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)
Catalog No. 50-131-1221 Supplier Chemglass Life Sciences Supplier No. CG3210100
Dual Temperature Controller Only, J-Kem, Apollo, Type "RTD" (-200C to 400C). J-Kem Scientific Apollo dual temperature controller features two independent temperature controllers in a single cabinet. Each independent channel has its own high power outlet, 100 hour timer to turn heating/cooling on or off at a user set time, and solid state over temperature protection circuit. Temperature ramping and serial communications are also provided with this unit.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 4" H x 8" W x 9" DPower: 10 Amps @ 120VAC, 1200 Watts MAX/Channel, 1800 Watts (Total)