Learn More
HARVARD BIOSCIENCE INC TEMP CONTROLLER DUAL CHNL
Catalog No. NC3404298
Encompass_Preferred
  • Dual-channel control for managing two heating elements.
  • Automatic and manual control modes.
  • Flexible temperature setting with internal and external adjustment options.
  • External control voltage scale is 100 mV/ºC.
  • Temperature range from ambient to 65 °C.
  • Loop speed options include fast, medium, slow.
  • Real-time thermistor output calibrated at 100 mV/ºC.
  • Display options for set temperature, T1 (control), T2 (monitor), and output voltage.
  • Manual output voltage range from 0 to +12 V DC.
  • Fault detection mechanisms protect against overheating.
Catalog No. NC3404298 Supplier HARVARD BIOSCIENCE INC Supplier No. 642401
  • Dual-channel control for managing two heating elements.
  • Automatic and manual control modes.
  • Flexible temperature setting with internal and external adjustment options.
  • External control voltage scale is 100 mV/ºC.
  • Temperature range from ambient to 65 °C.
  • Loop speed options include fast, medium, slow.
  • Real-time thermistor output calibrated at 100 mV/ºC.
  • Display options for set temperature, T1 (control), T2 (monitor), and output voltage.
  • Manual output voltage range from 0 to +12 V DC.
  • Fault detection mechanisms protect against overheating.