Learn More
Ace Glass, Inc. Temperature controller,economy,type J,0-800 degrees C
SDP
Catalog No. NC3246061
Encompass
Economy Model, compact, digital temperature controller. 16 segment ramp and soak, fuzzy logic, auto tune PID controller. One T, K, or J type sensor input. One 120V output. Temperature range varies by thermocouple type. Control accuracy -plus - minus 0.1 Degree C. Absolute accuracy of plus-minus 0.25 Degrees C of full range. Microprocessor based 1/32 DIN model for digital accuracy and reliability, Bright LED display. 120V, 15A, 50/60Hz. 3 year limited warranty.
Catalog No. NC3246061 Supplier Ace Glass, Inc. Supplier No. 1212514
Economy Model, compact, digital temperature controller. 16 segment ramp and soak, fuzzy logic, auto tune PID controller. One T, K, or J type sensor input. One 120V output. Temperature range varies by thermocouple type. Control accuracy -plus - minus 0.1 Degree C. Absolute accuracy of plus-minus 0.25 Degrees C of full range. Microprocessor based 1/32 DIN model for digital accuracy and reliability, Bright LED display. 120V, 15A, 50/60Hz. 3 year limited warranty.