Learn More
Chemglass Life Sciences J-Kem Temperature Controller Only, Model 210, Type "RTD" (-200°C to 400°C)
SDP
Catalog No. 501311216
Encompass_Preferred
J-Kem Temperature Controller Only, Model 210, Type "RTD" (-200C to 400C). The Model 210 has 1200 Watts of power which is sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates. Controller has the ability to display temperature without heating. The Model 210 controls temperature to within 0.1C of liquids from 1ul to 5L.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.25" H x 5.25" W x 7.25" DPower: 10 Amps @ 120VAC, 1200 Watts
Catalog No. 50-131-1216 Supplier Chemglass Life Sciences Supplier No. CG3200100
J-Kem Temperature Controller Only, Model 210, Type "RTD" (-200C to 400C). The Model 210 has 1200 Watts of power which is sufficient for heating mantles up to 5L in size along with most laboratory ovens and hot plates. Controller has the ability to display temperature without heating. The Model 210 controls temperature to within 0.1C of liquids from 1ul to 5L.PLEASE NOTE: THE TYPE T, J AND K CONTROLLERS ARE SUPPLIED COMPLETE WITH: 1/8 DIAMETER X 12 LONG PTFE COATED THERMOCOUPLE, 10FT COILED EXTENSION CORD, AND A 14/20 PTFE THERMOCOUPLE ADAPTER. THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.25" H x 5.25" W x 7.25" DPower: 10 Amps @ 120VAC, 1200 Watts