Learn More
Chemglass Life Sciences J-Kem Temperature Controller Only, Model 260/T, Type "RTD" (-200°C to 400°C)
SDP
Supplier:  Chemglass Life Sciences CG3206100
Encompass_Preferred
J-Kem Temperature Controller Only, Model 260/T, Type "RTD" (-200C to 400C). The Model 260/T disconnects all output power it the process temperature exceeds the entered temperature by a user-specified amount, or following recovery from a power failure. The outputs remain disconnected until manually reset. Over temperature condition is signalled by an indicator lamp and audible alarm. The Model 260/T has 1800 Watts of power which is sufficient for heating mantles up to 22L. Controller has the ability to display temperature without heating. The Model 260/T is also equipped with a100 hour digital timer to turn heating ON or OFF when time has expired.THE RTD UNIT IS THE CONTROLLER ONLY.Specifications:Dimensions: 3.25" H x 7.5" W x 9" DPower: 15 Amps @ 120VAC, 1800 Watts
Catalog No. 50-131-1219