Learn More
Electron Microscopy Sciences Corning Digital Stirring Hot Plate Temperature Controller
SDP
Catalog No. 501899842
Encompass_Preferred
This accessory works with all Corning hot plates, models PC-400, and PC-600, and hot plate/stirrers, models PC-420, and PC-620. Use to control or monitor temperature of hot plate. PTFE coated probe.
Use: PC-420
Catalog No. 50-189-9842 Supplier Electron Microscopy Sciences Supplier No. 7235121
This accessory works with all Corning hot plates, models PC-400, and PC-600, and hot plate/stirrers, models PC-420, and PC-620. Use to control or monitor temperature of hot plate. PTFE coated probe.
Use: PC-420