Learn More
Cole-Parmer Taylor 497J Metal Dial Thermometer, 12", -60 to 120F, 30% to 80% RH
Catalog No. NC0250682 Shop All Cole-Parmer Products
Encompass_Preferred
Giant, oversized face has bold, easy-to-read numerals. Made of metal for durability and is weather resistant. Thermometer easily mounts to walls or any other flat surface. Dual-scale thermometer reads in both C and F. Temperature scale has maximum contrast for ease of reading in low lighting conditions. Rectangle thermometers are 12" high and have a spirit filled glass tube.
Catalog No. NC0250682 Supplier Cole-Parmer Supplier No. 9000035
Giant, oversized face has bold, easy-to-read numerals. Made of metal for durability and is weather resistant. Thermometer easily mounts to walls or any other flat surface. Dual-scale thermometer reads in both C and F. Temperature scale has maximum contrast for ease of reading in low lighting conditions. Rectangle thermometers are 12" high and have a spirit filled glass tube.