Learn More
Research Products International Corp Humidity/Temperature Monitor with Product Certification
SDP
Catalog No. 50550387
Encompass
Simultaneously monitors both temperature and humidity on an extra large 3 3/16 x 2 1/2 inch dual display which can be read from across a room. Monitor humidity and temperature in labs, chemical storage areas, clean rooms, stock rooms, warehouses, factories, greenhouses, and outdoors. Recall min/max temperature and min/max humidity readings over any time period or have it automatically reset itself every 24 hours. Stand or wall mount.
Comes with 1 AAA battery which lasts a full year and N.I.S.T. traceable certificate of compliance.
Catalog No. 50-550-387 Supplier Research Products International Corp Supplier No. 800016C
Simultaneously monitors both temperature and humidity on an extra large 3 3/16 x 2 1/2 inch dual display which can be read from across a room. Monitor humidity and temperature in labs, chemical storage areas, clean rooms, stock rooms, warehouses, factories, greenhouses, and outdoors. Recall min/max temperature and min/max humidity readings over any time period or have it automatically reset itself every 24 hours. Stand or wall mount.
Comes with 1 AAA battery which lasts a full year and N.I.S.T. traceable certificate of compliance.