Learn More
Cole-Parmer Irreversible 8-Point Vertical Temperature Label, 160-230F/71-110C; 25/Pk
Catalog No. NC9448004 Shop All Cole-Parmer Products
Encompass_Preferred
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8F below 212F and ±1F above 212F. Shelf life: 1 year after date of purchase.
Catalog No. NC9448004 Supplier Cole-Parmer Supplier No. 0806822
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8F below 212F and ±1F above 212F. Shelf life: 1 year after date of purchase.