Learn More
Cole-Parmer Irreversible 10 Point Vertical Temperature Label, 171-261F/77-127C; 10/Pk
Catalog No. 501961540 Shop All Cole-Parmer Products
Encompass_Preferred
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1C from 29 to 99C, ±1.5C from 100 to 154C, and ±1% + 1C from 160 to 280C. Shelf life: one year after date of purchase.
Catalog No. 50-196-1540 Supplier Cole-Parmer Supplier No. 0903506
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1C from 29 to 99C, ±1.5C from 100 to 154C, and ±1% + 1C from 160 to 280C. Shelf life: one year after date of purchase.