Learn More
Research Products International Corp Temp Monitor, Dual Sensor
SDP
Catalog No. 501978366
Encompass
Simultaneously displays internal and external temperature on a 3 3/16 x 2 1/2 inch LCD, large enough to be read across a room. Useful as an indoor/outdoor thermometer, in incubators or anywhere simultaneous internal and external readings are required. Has min/max temperature memory. Can be wall mounted or freestanding with easel back. Recall min/max readings over any time period or have it automatically reset itself every 24 hours. The outdoor sensor is mounted on a cord over 4 feet long.
Catalog No. 50-197-8366 Supplier Research Products International Corp Supplier No. 800015
Simultaneously displays internal and external temperature on a 3 3/16 x 2 1/2 inch LCD, large enough to be read across a room. Useful as an indoor/outdoor thermometer, in incubators or anywhere simultaneous internal and external readings are required. Has min/max temperature memory. Can be wall mounted or freestanding with easel back. Recall min/max readings over any time period or have it automatically reset itself every 24 hours. The outdoor sensor is mounted on a cord over 4 feet long.