Learn More
Lascar Electronics Inc Temperature monitor with email alerts
SDP
Catalog No. NC1856634
Encompass_Preferred
Wireless Alert TP is a battery-powered temperature sensor that sends a warning email if a user-set level is reached. Simply select your temperature limits, add an email address and connect to your Wi-Fi network using the Wireless Alert mobile app.
Catalog No. NC1856634 Supplier Lascar Electronics Inc Supplier No. WIRELESS ALERT TP
Wireless Alert TP is a battery-powered temperature sensor that sends a warning email if a user-set level is reached. Simply select your temperature limits, add an email address and connect to your Wi-Fi network using the Wireless Alert mobile app.