Learn More
USA Scientific Inc TempPlate non-skirted 96-well PCR plate, low profile, natural
Catalog No. NC9420961
Encompass
TempPlate non-skirted polypropylene 0.1 mL 96-well PCR plate, low profile, H12 notch, natural. Two/sleeve, 10/box.
Catalog No. NC9420961 Supplier USA Scientific Inc Supplier No. 14029500
TempPlate non-skirted polypropylene 0.1 mL 96-well PCR plate, low profile, H12 notch, natural. Two/sleeve, 10/box.