Learn More
Physitemp TEMP PROBE
Catalog No. NC9034673
Encompass_Preferred
Rectal probe for rats, For fast intermittent measurements, Smooth ball tip, Stainless steel shaft, 1 in. L, 0.59 in. dia., tip dia. 0.125 in., 5' lead, Max Temp.: 125degreeC, Time constant 0.8 sec., Not Isolated
Catalog No. NC9034673 Supplier Physitemp Supplier No. RET2
Rectal probe for rats, For fast intermittent measurements, Smooth ball tip, Stainless steel shaft, 1 in. L, 0.59 in. dia., tip dia. 0.125 in., 5' lead, Max Temp.: 125degreeC, Time constant 0.8 sec., Not Isolated