Learn More
Lascar Electronics Inc Glycol Probe for EasyLog Data Loggers
SDP
Catalog No. NC1587414
Encompass_Preferred
High accuracy, 3 metre long thermistor type A probe in sealed Glycol filled bottle for vaccine monitoring.Works with the following products: EL-21CFR-TP-LCD+, EL-USB-TP-LCD+, EL-GFX-DTP+, EL-WIFI-TP+, EL-WIFI-DTP+.
Catalog No. NC1587414 Supplier Lascar Electronics Inc Supplier No. ELPVAC
High accuracy, 3 metre long thermistor type A probe in sealed Glycol filled bottle for vaccine monitoring.Works with the following products: EL-21CFR-TP-LCD+, EL-USB-TP-LCD+, EL-GFX-DTP+, EL-WIFI-TP+, EL-WIFI-DTP+.