Learn More
Cole-Parmer Digi-Sense Type K High-Temperature Fle x ible Thermocouple Probe; 12" L
Supplier:  Cole-Parmer 0851696
Encompass_Preferred
High-temperature thermocouple probes designed for multiple bends are ideal for any environment. Bend probes around pipe diameters as small as twice the sheath diameter to easily determine your pipe temperature. Ideal for industrial environments where fast accurate readings are required. Response Time: 85 seconds or greater. Type K and J probes feature ICONEL 600 sheaths. Type T probes feature 316 stainless steel sheaths.
Catalog No. NC9689471