Learn More
Lascar Electronics Inc Dual Channel Temp, RH & Dew Point Data Logger with External Probe, Graphic Screen and Audible Alarm
SDP
Catalog No. NC1856720
Encompass_Preferred
This dual channel data logger measures and stores over 250,000 temperature and relative humidity readings using up to two external temperature & humidity probes*. It has user-programmable, audible alarm thresholds with confidence/alarm LEDs. Its high contrast, dot-matrix LCD with three face-buttons for navigation gives real-time trend analysis, data summaries and the ability to start, stop and restart the data logger without connecting to a PC. EasyLog USB software enables quick configuration and data download via the USB port. Supplied with two lithium batteries, which can typically allow logging for up to 1 year. It can also be powered from USB.Logging rates between 10 seconds and 1 hourStores over 250,000 readings per channel for temperature and humidityUser triggered logging modeUp to two channels supported (supplied with one probe)*only one probe is included with the device.
Catalog No. NC1856720 Supplier Lascar Electronics Inc Supplier No. ELGFXD2
This dual channel data logger measures and stores over 250,000 temperature and relative humidity readings using up to two external temperature & humidity probes*. It has user-programmable, audible alarm thresholds with confidence/alarm LEDs. Its high contrast, dot-matrix LCD with three face-buttons for navigation gives real-time trend analysis, data summaries and the ability to start, stop and restart the data logger without connecting to a PC. EasyLog USB software enables quick configuration and data download via the USB port. Supplied with two lithium batteries, which can typically allow logging for up to 1 year. It can also be powered from USB.Logging rates between 10 seconds and 1 hourStores over 250,000 readings per channel for temperature and humidityUser triggered logging modeUp to two channels supported (supplied with one probe)*only one probe is included with the device.