Learn More
Lascar Electronics Inc Temperature and humidty data logger with display
SDP
Supplier:  Lascar Electronics Inc ELSIE2+
Encompass_Preferred
Monitor the environment you live and work in with the EasyLog EL-SIE-2+. Advanced onboard software means there is nothing to install on your PC or Mac – simply connect the logger with a USB cable and use your web browser to configure the device and view, analyse and download your data. It really couldn’t be easier.The bright and clear display shows current readings and device status at a glance, and the product is supplied with a bracket for secure wall mounting. EL-SIE-2+ uses standard AAA batteries and typical battery life is over 1 year.The EL-SIE-2+ is a high-accuracy model version and measures temperature and relative humidity. Other models that measure temperature only, or temperature, humidity, and pressure, are available. Standard and high accuracy versions of these are also available. Key markets include facilities and building management, logistics, warehouses and storage, and clean rooms and other pressure-sensitive environments.
Catalog No. NC1856665