Learn More
New England Biolabs, Inc. Template Switching RT Enzyme Mix – 100 reactions
SDP
Catalog No. 501959376
Encompass
The reverse transcriptase (RT) in the Template Switching RT Enzyme Mix adds a few non-templated nucleotides after it reaches the 5' end of the RNA template. These non-templated nucleotides can anneal to a template switching oligo (TSO) with a known sequence, prompting the reverse transcriptase to switch template from RNA to the TSO. The resulting cDNA contains a known sequence (complementary to the sequence of the TSO) attached to the 3' end. This feature can be utilized in a variety of downstream applications, such as cDNA amplification, 5' RACE (rapid amplification of cDNA ends), and 2nd strand cDNA synthesis.
Catalog No. 50-195-9376 Supplier New England Biolabs, Inc. Supplier No. M0466L
The reverse transcriptase (RT) in the Template Switching RT Enzyme Mix adds a few non-templated nucleotides after it reaches the 5' end of the RNA template. These non-templated nucleotides can anneal to a template switching oligo (TSO) with a known sequence, prompting the reverse transcriptase to switch template from RNA to the TSO. The resulting cDNA contains a known sequence (complementary to the sequence of the TSO) attached to the 3' end. This feature can be utilized in a variety of downstream applications, such as cDNA amplification, 5' RACE (rapid amplification of cDNA ends), and 2nd strand cDNA synthesis.