Learn More
Pharmacal Research Laboratories Temp-Tapes® 180  , 24pk
SDP
Catalog No. NC9333684
Encompass
Temp-Tape® 180o is a non-reversible, cage surface temperature-recording label. The light gray background will turn black seconds after being exposed to 180o water. Accuracy of this tape is 99%. The exposed recording tape may be removed and filed for future reference. We recommend the use of this tape for verifying every load in all automatic cage washing equipment.
Catalog No. NC9333684 Supplier Pharmacal Research Laboratories Supplier No. 99054
Temp-Tape® 180o is a non-reversible, cage surface temperature-recording label. The light gray background will turn black seconds after being exposed to 180o water. Accuracy of this tape is 99%. The exposed recording tape may be removed and filed for future reference. We recommend the use of this tape for verifying every load in all automatic cage washing equipment.