Learn More
HANNA INSTRUMENTS INC Multiparameter pH/ORP/EC/TDS/Salinity/DO/Pressure/ Temperature Waterproof Meter
SDP
Catalog No. NC1883639
Encompass
The HI98194 is a rugged, waterproof multiparameter meter that monitors up to 12 different key water quality parameters.A backlit, graphic LCD provides easy to read resolution even in low-lit areas and a combination of dedicated and soft keys allows easy operation. The powerful logging capability and digital probe make this multiparameter water quality meter a great tool.Comes as a complete system, allowing you to start taking measurements as soon as you receive it.CAL Check™ electrode diagnostics system alerts you to potential calibration problems so that you know your results are trustworthy every time.Great for environmental and industrial professionals.
Catalog No. NC1883639 Supplier HANNA INSTRUMENTS INC Supplier No. HI98194
The HI98194 is a rugged, waterproof multiparameter meter that monitors up to 12 different key water quality parameters.A backlit, graphic LCD provides easy to read resolution even in low-lit areas and a combination of dedicated and soft keys allows easy operation. The powerful logging capability and digital probe make this multiparameter water quality meter a great tool.Comes as a complete system, allowing you to start taking measurements as soon as you receive it.CAL Check™ electrode diagnostics system alerts you to potential calibration problems so that you know your results are trustworthy every time.Great for environmental and industrial professionals.