Learn More
USA Scientific Inc 0.2 mL PCR 8-tube FLEX-FREE strip, attached optical cap strip, mixed
Catalog No. NC1879251
Encompass
0.2 mL TempAssure PCR 8-tube flex-free hinge-strips, color tubes with attached optically clear 8-cap strips. 125 strips of tubes with caps per pack.
Catalog No. NC1879251 Supplier USA Scientific Inc Supplier No. 14021808
0.2 mL TempAssure PCR 8-tube flex-free hinge-strips, color tubes with attached optically clear 8-cap strips. 125 strips of tubes with caps per pack.