Learn More
BINDER™ Analog Output
SDP
Catalog No. 08112019 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MK
Catalog No. For Use With (Equipment)
08-112-019 BINDER Chambers Series MK
1 options
Catalog No. 08-112-019 Supplier BINDER™ Supplier No. 80121084
Add to Cart
Add to Cart
For Use With: All MK Series models
  • Output not adjustable
  • 4 to 20 mA
Specifications
Description Analog output 4-20 mA - for temperature values (output not adjustable)
For Use With (Equipment) BINDER Chambers Series MK
Material Plastic
Type Analog Output
For Use With (Application) Temperature