Learn More
BINDER™ Temperature Analog Output
SDP
Catalog No. 08111583 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-583 Plastic
1 options
Catalog No. 08-111-583 Supplier BINDER™ Supplier No. 80121625
Add to Cart
Add to Cart
For Use With: FED 56, FED 115, FED 260, FED 720
  • Output not adjustable
  • 4 to 20 mA
Specifications
Description Analog output 4-20 mA - for temperature values (output not adjustable)
For Use With (Equipment) BINDER Chambers FED 56, FED 115, FED 260, FED 720
Material Plastic
Type Analog Output
For Use With (Application) Temperature