Learn More
BINDER™ Temperature Analog Output
SDP
Catalog No. 08111572 Shop All BINDER Inc. Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
08-111-572 Plastic
1 options
Catalog No. 08-111-572 Supplier BINDER™ Supplier No. 80120430
Add to Cart
Add to Cart
For Use With: BINDER units ED 23, ED 400
  • Output not adjustable
  • 4 to 20 mA
Specifications
Description Analog output 4-20 mA - for temperature values (output not adjustable)
For Use With (Equipment) BINDER Chambers ED 23, ED 400
Material Plastic
Type Analog Output
For Use With (Application) Temperature