Description
- Output not adjustable
- 4 to 20 mA
Specifications
Specifications
| Description | Analog output 4-20 mA - for temperature values (output not adjustable) |
| For Use With (Equipment) | BINDER Chambers BD 56, BD 115, BD 260, BD 720 |
| Material | Plastic |
| Type | Analog Output |
| For Use With (Application) | Temperature |